missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-N-methyl-L-glutamic acid 5-tert-butyl ester, 98%

Brand:  Alfa Aesar™ H63814.MD

Additional Details : CAS Number : 200616-40-6 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15448536

  • 2790.92 SEK / 250mg


Estimated Shipment date
from Supplier 23-01-2019
Add to basket

Specifications

Chemical Identifiers

200616-40-6
439.508
FVUASVBQADLDRO-NRFANRHFSA-N
40428349
CC(C)(C)OC(=O)CCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C25H29NO6
MFCD00237028
fmoc-n-me-glu otbu-oh, fmoc-n-methyl-l-glutamic acid 5-tert-butyl ester, 2s-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-5-oxopentanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-5-tert-butoxy-5-oxopentanoic acid, ambotzfaa1397, fmoc-n-me-glu otbu-oh, nalpha-fmoc-nalpha-methyl-l-glutamic acid gamma-tert-butyl ester, n-9-fluorenylmethyloxycarbonyl-n-methyl-l-glutamic acid 5-tert-butyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-glutamic-acid-gamma-t-butyl ester, 2s-5-tert-butoxy-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-5-oxopentanoic acid
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid

Specifications

N-Fmoc-N-methyl-L-glutamic acid 5-tert-butyl ester
C25H29NO6
fmoc-n-me-glu otbu-oh, fmoc-n-methyl-l-glutamic acid 5-tert-butyl ester, 2s-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-5-oxopentanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-5-tert-butoxy-5-oxopentanoic acid, ambotzfaa1397, fmoc-n-me-glu otbu-oh, nalpha-fmoc-nalpha-methyl-l-glutamic acid gamma-tert-butyl ester, n-9-fluorenylmethyloxycarbonyl-n-methyl-l-glutamic acid 5-tert-butyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-glutamic-acid-gamma-t-butyl ester, 2s-5-tert-butoxy-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
439.508
439.5
98%
200616-40-6
MFCD00237028
FVUASVBQADLDRO-NRFANRHFSA-N
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
40428349
Solid
250mg
Videos
SDS