missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-norleucine, 96%, Thermo Scientific Chemicals

Catalog Number p-7055416
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
112883-42-8
C22H25NO4
367.45
MFCD00235878
RZZXDYZWHUAOEK-GNLPSFAGNA-N
fmoc-n-me-nle-oh, fmoc-n-methyl-l-norleucine, fmoc-nalpha-methyl-l-norleucine, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-norleucine, ambotzfaa6460, fmoc-n-me-nle-oh hplc
7019522
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]hexanoic acid
CCCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
112883-42-8
C22H25NO4
367.45
MFCD00235878
RZZXDYZWHUAOEK-GNLPSFAGNA-N
fmoc-n-me-nle-oh, fmoc-n-methyl-l-norleucine, fmoc-nalpha-methyl-l-norleucine, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-norleucine, ambotzfaa6460, fmoc-n-me-nle-oh hplc
7019522
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]hexanoic acid
CCCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 367.44
Percent Purity 96%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Fmoc-N-methyl-L-norleucine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'