Läs mer
N-Formyl Bisoprolol, TRC
Produktkod. 30407944
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407944 50 mg
30405120 10 mg
30406674 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30407944 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF69730350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1447715-45-8
Molekylformel C19 H31 N O5
Molekylvikt (g/mol) 353.45
Synonym N-[2-Hydroxy-3-[4-[[2-(1-methylethoxy)ethoxy]methyl]phenoxy]propyl]-N-(1-methylethyl)formamide
IUPAC-namn N-[2-hydroxy-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propyl]-N-propan-2-ylformamide
LEDER CC(C)OCCOCc1ccc(OCC(O)CN(C=O)C(C)C)cc1
Specifikationer
Kemiskt namn eller material N-Formylbisoprolol
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C19H31NO5/c1-15(2)20(14-21)11-18(22)13-25-19-7-5-17(6-8-19)12-23-9-10-24-16(3)4/h5-8,14-16,18,22H,9-13H2,1-4H3
Formel vikt 353.2202
Procent renhet >95%