Läs mer
N-Formyl-L-leucine (3S,4R,6S)-3-Hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl Ester, TRC
Produktkod. 30010954
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010954 2.5 mg 2.5mg
30007573 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010954 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF7005702.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 130793-27-0
Molekylformel C29 H53 N O5
Molekylvikt (g/mol) 495.73
Synonym L-Leucine, N-formyl-, (3S,4R,6S)-3-hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl ester, L-Leucine, N-formyl-, 3-hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl ester, [3S-(3α,4β,6β)]-, Orlistat Related Compound D (USP)
IUPAC-namn [(3S,4R,6S)-3-hexyl-2-oxo-6-undecyloxan-4-yl] (2S)-2-formamido-4-methylpentanoate
LEDER CCCCCCCCCCC[C@H]1C[C@@H](OC(=O)[C@H](CC(C)C)NC=O)[C@H](CCCCCC)C(=O)O1
Specifikationer
Kemiskt namn eller material N-formyl-L-leucine (3S,4R,6S)-3-hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl ester
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C29H53NO5/c1-5-7-9-11-12-13-14-15-16-18-24-21-27(25(28(32)34-24)19-17-10-8-6-2)35-29(33)26(30-22-31)20-23(3)4/h22-27H,5-21H2,1-4H3,(H,30,31)/t24-,25-,26-,27+/m0/s1
Kvantitet 2.5 mg
Formel vikt 495.3924
Procent renhet >95%