Läs mer
N-Formyl-Met-Leu-Phe, TRC
Produktkod. 30402411
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30402411 5 mg
1 options
Produktkod. 30402411 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF7007135MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Formyl-Met-Leu-Phe
CAS 59880-97-6
Molekylformel C21H31N3O5S
Kvantitet 5 mg
Synonym Chemotactic Peptide
InChI formel InChI=1S/C21H31N3O5S/c1-14(2)11-17(23-19(26)16(22-13-25)9-10-30-3)20(27)24-18(21(28)29)12-15-7-5-4-6-8-15/h4-8,13-14,16-18H,9-12H2,1-3H3,(H,22,25)(H,23,26)(H,24,27)(H,28,29)/t16-,17-,18-/m0/s1
LEDER CSCC[C@H](NC=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc1ccccc1)C(=O)O
Molekylvikt (g/mol) 437.55
Formel vikt 437.1984