Läs mer
N-Formylcytisine, TRC
Produktkod. 30006169
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006169 10 mg
30005352 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006169 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF69897010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53007-06-0
Molekylformel C12 H14 N2 O2
Molekylvikt (g/mol) 218.25
Synonym 1,5-Methano-2H-pyrido[1,2-a][1,5]diazocine-3(4H)-carboxaldehyde, 1,5,6,8-tetrahydro-8-oxo-, (1R,5S)-, 1,5-Methano-2H-pyrido[1,2-a][1,5]diazocine-3(4H)-carboxaldehyde, 1,5,6,8-tetrahydro-8-oxo-, (1R)-, (1R,5S)-1,5,6,8-Tetrahydro-8-oxo-1,5-methano-2H-pyrido[1,2-a][1,5]diazocine-3(4H)-carboxaldehyde, (-)-N-Formylcytisine, N-Formylcytisine
LEDER O=CN1C[C@@H]2C[C@H](C1)C3=CC=CC(=O)N3C2
Specifikationer
Kemiskt namn eller material N-Formylcytisine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C12H14N2O2/c15-8-13-5-9-4-10(7-13)11-2-1-3-12(16)14(11)6-9/h1-3,8-10H,4-7H2/t9-,10+/m0/s1
Formel vikt 218.1055
Procent renhet >95%