Läs mer
N-Glycolyl Neuraminic Acid, TRC
Product Code. 30004659
Change view
Click to view available options
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30004659 25 mg
30012716 10 mg
30011444 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004659 Supplier Toronto Research Chemicals Supplier No. TRCG65000025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1113-83-3
Molekylformel C11 H19 N O10
Molekylvikt (g/mol) 325.27
IUPAC-namn (4S,5R,6R,7S,8R)-4,6,7,8,9-pentahydroxy-5-[(2-hydroxyacetyl)amino]-2-oxononanoic acid
LEDER OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](NC(=O)CO)[C@@H](O)CC(=O)C(=O)O
Specifications
Kemiskt namn eller material N-Glycolylneuraminic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C11H19NO10/c13-2-6(17)9(19)10(20)8(12-7(18)3-14)4(15)1-5(16)11(21)22/h4,6,8-10,13-15,17,19-20H,1-3H2,(H,12,18)(H,21,22)/t4-,6+,8+,9+,10+/m0/s1
Formel vikt 325.1009
Procent renhet >95%