missing translation for 'onlineSavingsMsg'
Learn More

n-Hexanal dimethylacetal, 98+%, Thermo Scientific™

Catalog Number p-3593477
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1599-47-9
C8H18O2
146.23
MFCD00036508
ZQUJUCJLDXTKKW-UHFFFAOYSA-N
74137
1,1-dimethoxyhexane
CCCCCC(OC)OC
This item is not returnable. View return policy
1599-47-9
C8H18O2
146.23
MFCD00036508
ZQUJUCJLDXTKKW-UHFFFAOYSA-N
74137
1,1-dimethoxyhexane
CCCCCC(OC)OC
CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Density 0.8470g/mL
Boiling Point 145°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(OCH3)2
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.847
Synonym hexanal dimethyl acetal, hexane, 1,1-dimethoxy, aldehyde c-6 dimethyl acetal, acmc-20akrj, 1,1-dimethoxy-hexane, hexane,1,1-dimethoxy, hexaldehyde dimethyl acetal, 1,1-dimethoxyhexane
Solubility Information Solubility in water: practically insoluble
Formula Weight 146.23
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material n-Hexanal dimethylacetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Hexanal dimethylacetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'