missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecane, MP Biomedicals™

Catalog Number p-4405811
Click to view available options
Quantity:
0.5 mL
100 mL
250 mL
1 L
This item is not returnable. View return policy
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
CHEBI:29021
hexane
CCCCCC
This item is not returnable. View return policy
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
CHEBI:29021
hexane
CCCCCC

n-Hexadecane ≥99%, Liquid

  • Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP. Hexadecane (n-Hexadecane) is an aliphatic long-chain hydrocarbon. It is a reference fuel with the cetane number 100 and is employed in determining the cetane number of diesel fuels. The mechanism of the oxidation of hexadecane in the gas-phase has been described. The density, speed of sound, adiabatic compressibility and compressed liquid density of hexadecane have been determined. The performance of platinum/zeolite catalysts for the hydroisomerization of n-hexadecane has been assessed. The thermal degradation of n-hexadecane forms n-alkanes and 1-alkenes as key products.
Melting Point -95°C (Literature)
Color 1 (APHA)
Density 0.66 at 20°C (water = 1); 0.659g/mL at 25°C(Lit.)
Boiling Point 69°C (Literature)
Quantity 0.5 mL
Physical Form Liquid
Refractive Index n20/D 1.375(Lit.)
Assay Percent Range 99.9%
Residue after Evaporation <0 ppm
Specific Gravity 0.674
Vapor Pressure 124 mm Hg at 20°C 150 mm Hg at 24.8°C; 256 mmHg ( 37.7°C), 5.2 psi ( 37.7°C), ∽132 mmHg ( 20°C)(Lit.)
UN Number UN1208
Beilstein 1730733
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Solubility Information Soluble in water.
Formula Weight 86.18
Chemical Name or Material n-Hexane, For ACS analysis
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'