Läs mer
N-(Hexanoyloxy)succinimide, TRC
Produktkod. 30403347
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30403347 100 mg
1 options
Produktkod. 30403347 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH281215100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(Hexanoyloxy)succinimide
CAS 22102-92-7
Molekylformel C10 H15 N O4
Kvantitet 100 mg
Synonym 1-[(1-Oxohexyl)oxy]-2,5-pyrrolidinedione, Hexanoic Acid 2,5-Dioxo-1-pyrrolidinyl Ester
InChI formel InChI=1S/C10H15NO4/c1-2-3-4-5-10(14)15-11-8(12)6-7-9(11)13/h2-7H2,1H3
LEDER CCCCCC(=O)ON1C(=O)CCC1=O
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) hexanoate
Molekylvikt (g/mol) 213.23
Formel vikt 213.1001