missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N'-Hydroxy-2-(trifluoromethoxy)benzenecarboximidamide, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | N'-hydroxy-2-(trifluoromethoxy)benzene-1-carboximidamide |
Formula Weight | 220.15 |
Percent Purity | 95% |
Chemical Name or Material | N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'