Learn More
n-Hydroxy-butyramidine, ≥97%, Thermo Scientific™
Catalog Number p-3076404
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
27620-10-6
C4H10N2O
102.137
MFCD06200896
OPENCMFJZQABIY-UHFFFAOYSA-N
n-hydroxybutyramidine, 1z-n'-hydroxybutanimidamide, n-hydroxybutyrimidamide, z-n'-hydroxybutyrimidamid e, butanimidamide, n-hydroxy, z-n'-hydroxybutimidamide, butanimidamide,n-hydroxy, e-n'-hydroxybutanimidamide, 1-1h-benzo d imidazol-2-yl-3-methyl-4-oxo-1,4-di
9582833
N'-hydroxybutanimidamide
CCCC(=NO)N
27620-10-6
C4H10N2O
102.137
MFCD06200896
OPENCMFJZQABIY-UHFFFAOYSA-N
n-hydroxybutyramidine, 1z-n'-hydroxybutanimidamide, n-hydroxybutyrimidamide, z-n'-hydroxybutyrimidamid e, butanimidamide, n-hydroxy, z-n'-hydroxybutimidamide, butanimidamide,n-hydroxy, e-n'-hydroxybutanimidamide, 1-1h-benzo d imidazol-2-yl-3-methyl-4-oxo-1,4-di
9582833
N'-hydroxybutanimidamide
CCCC(=NO)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 102.14
Percent Purity 97%
Chemical Name or Material N-hydroxy-butyramidine