Läs mer
N-Hydroxy Sertraline, TRC
Produktkod. 30408087
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30408087 10 mg
30400261 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30408087 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95373010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 124345-07-9
Molekylformel C17 H17 Cl2 N O
Molekylvikt (g/mol) 322.23
Synonym (1S-cis)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-hydroxy-N-methyl-1-naphthalenamine, N-Hydroxysertraline
IUPAC-namn [(2R,5S)-5-acetyloxy-3-benzoyloxyoxolan-2-yl]methyl benzoate
LEDER N(C)(O)[C@@H]1C=2C([C@@H](CC1)C3=CC(Cl)=C(Cl)C=C3)=CC=CC2
Specifikationer
Kemiskt namn eller material N-Hydroxy Sertraline
Kvantitet 10 mg
InChI formel InChI=1S/C17H17Cl2NO/c1-20(21)17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11/h2-6,8,10,12,17,21H,7,9H2,1H3/t12-,17-/m0/s1
Formel vikt 321.0687