Läs mer
N-Hydroxy Sulfamethoxazole, TRC
Produktkod. 30134092
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134092 25 mg
30010876 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134092 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95380025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 114438-33-4
Molekylformel C10 H11 N3 O4 S
Molekylvikt (g/mol) 269.28
Synonym 4-(Hydroxyamino)-N-(5-methyl-3-isoxazolyl)-benzenesulfonamide, Sulfamethoxazole N4-Hydroxylamine, Sulfamethoxazole Hydroxylamine, NSC 665547;
IUPAC-namn 4-(hydroxyamino)-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
LEDER Cc1onc(NS(=O)(=O)c2ccc(NO)cc2)c1
Specifikationer
Kemiskt namn eller material N-Hydroxy Sulfamethoxazole
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C10H11N3O4S/c1-7-6-10(12-17-7)13-18(15,16)9-4-2-8(11-14)3-5-9/h2-6,11,14H,1H3,(H,12,13)
Formel vikt 269.047
Procent renhet >95%