Läs mer
N-(Hydroxymethyl)nicotinamide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(Hydroxymethyl)nicotinamide |
| CAS | 3569-99-1 |
| Molekylformel | C7 H8 N2 O2 |
| Kvantitet | 25 g |
| Synonym | 3-Pyridinecarboxamide, N-(hydroxymethyl)-, Nicotinamide, N-(hydroxymethyl)- (6CI,7CI,8CI), N-(Hydroxymethyl)-3-pyridinecarboxamide, 3-Pyridinecarboxylic acid N-hydroxymethylamide, 3-Pyridinecarboxylic acid hydroxymethylamide, Bilamid, Bilizorin, Bilocid, Biloide, Biobilan, C 1094, Cholamid, Cholamide, Cholamide (pyridine derivative), Choligen, Coloton, Felosan, Isochol, N-(Hydroxymethyl)nicotinamide, Nicodin, Nicodine, Nicoform, Nicometamide, Nifoform, Nikodin, Nikoform, Nikomethamide |
| InChI formel | InChI=1S/C7H8N2O2/c10-5-9-7(11)6-2-1-3-8-4-6/h1-4,10H,5H2,(H,9,11) |
| LEDER | OCNC(=O)c1cccnc1 |
| IUPAC-namn | N-(hydroxymethyl)pyridine-3-carboxamide |
| Molekylvikt (g/mol) | 152.15 |
| Formel vikt | 152.0586 |