Läs mer
N(Im)-Ethoxycarbonyl-S-ethoxycarbonyl L-Ergothioneine Methyl Ester Iodide, TRC
Produktkod. 30377535
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30377535 50 mg
1 options
Produktkod. 30377535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE89047550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N(Im)-Ethoxycarbonyl-S-ethoxycarbonyl L-Ergothioneine Methyl Ester Iodide
CAS 162240-57-5
Molekylformel C16H26IN3O6S
Kvantitet 50 mg
Synonym (S)-1-(Ethoxycarbonyl)-2-[(ethoxycarbonyl)thio]-alpha-(methoxycarbonyl)-N,N,N-trimethyl-1H-imidazole-4-ethanaminium Iodide
InChI formel InChI=1S/C16H26N3O6S.HI/c1-7-24-15(21)18-10-11(17-14(18)26-16(22)25-8-2)9-12(13(20)23-6)19(3,4)5;/h10,12H,7-9H2,1-6H3;1H/q+1;/p-1/t12-;/m0./s1
LEDER [I-].CCOC(=O)Sc1nc(C[C@@H](C(=O)OC)[N+](C)(C)C)cn1C(=O)OCC
IUPAC-namn [(2S)-3-(1-ethoxycarbonyl-2-ethoxycarbonylsulfanylimidazol-4-yl)-1-methoxy-1-oxopropan-2-yl]-trimethylazanium;iodide
Molekylvikt (g/mol) 515.36
Formel vikt 515.0587