Läs mer
N-(Imidazolidin-2-ylidene)-4,6-dimethoxy-2-methylpyrimidin-5-amine (4-Methoxymoxonidine), Mikromol™
Produktkod. 30040401
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040401 100 mg
18083170 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30040401 Leverantör Mikromol Leverantörsnummer MM1278.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(Imidazolidin-2-ylidene)-4,6-dimethoxy-2-methylpyrimidin-5-amine (4-Methoxymoxonidine)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(Imidazolidin-2-ylidene)-4,6-dimethoxy-2-methylpyrimidin-5-amine (4-Methoxymoxonidine)
CAS 75439-01-9
Analyt- eller komponentnamn N-(Imidazolidin-2-ylidene)-4,6-dimethoxy-2-methylpyrimidin-5-amine (4-Methoxymoxonidine)
Frakt skick Room Temperature
Molekylformel C10 H15 N5 O2
Certifikat och efterlevnad ISO 9001
Synonym N-(Imidazolidin-2-ylidene)-4,6-dimethoxy-2-methylpyrimidin-5-amine, Moxonidine Imp. B (EP), 4-Methoxymoxonidine, Moxonidine Impurity B
InChI formel InChI=1S/C10H15N5O2/c1-6-13-8(16-2)7(9(14-6)17-3)15-10-11-4-5-12-10/h4-5H2,1-3H3,(H2,11,12,15)
LEDER COc1nc(C)nc(OC)c1N=C2NCCN2
IUPAC-namn N-(4,6-dimethoxy-2-methylpyrimidin-5-yl)imidazolidin-2-imine
Molekylvikt (g/mol) 237.26
Kvantitet 100 mg
Formel vikt 237.1226 g/mol