missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Isopropylacrylamide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 60°C to 63°C |
Boiling Point | 89°C to 92°C (2 mmHg) |
Quantity | 25 g |
Beilstein | 1744926 |
Solubility Information | Slightly soluble in water. |
IUPAC Name | N-(propan-2-yl)prop-2-enamide |
Formula Weight | 113.16 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | N-Isopropylacrylamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'