Läs mer
N-Isopropylnortropine Methobromide (Impurity), TRC
Produktkod. 30406102
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30406102 100 mg
1 options
Produktkod. 30406102 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI872685100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R,3r,5S,8r)-3-hydroxy-8-methyl-8-(1-methylethyl)-8-azoniabicyclo(3.2.1)octane
CAS 58005-18-8
Molekylformel C11 H22 N O . Br
Kvantitet 100 mg
Synonym 8-Azoniabicyclo[3.2.1]octane, 3-hydroxy-8-methyl-8-(1-methylethyl)-, bromide (1:1), (3-endo,8-syn)-, 8-Azoniabicyclo[3.2.1]octane, 3-hydroxy-8-methyl-8-(1-methylethyl)-, bromide, (endo,syn)- (9CI), (8R)-3α-Hydroxy-8-isopropyl-1αH,5αH-tropanium bromide, N-Isopropylnortropine methobromide, (1R,3r,5S,8r)-3-Hydroxy-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane Bromide
InChI formel InChI=1S/C11H22NO.BrH/c1-8(2)12(3)9-4-5-10(12)7-11(13)6-9;/h8-11,13H,4-7H2,1-3H3;1H/q+1;/p-1/t9-,10+,11+,12+;
LEDER [Br-].CC(C)[N@@+]1(C)[C@@H]2CC[C@H]1C[C@@H](O)C2
IUPAC-namn (1R,5S)-8-methyl-8-propan-2-yl-8-azoniabicyclo[3.2.1]octan-3-ol;bromide
Molekylvikt (g/mol) 264.2
Formel vikt 263.0885