Läs mer
N-[(-)-Jasmonoyl]-(L)-isoleucine, TRC
Produktkod. 30007710
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007710 50 mg
30006247 25 mg
30005602 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007710 Leverantör Toronto Research Chemicals Leverantörsnummer TRCJ21056350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 120330-93-0
Molekylformel C18 H29 N O4
Molekylvikt (g/mol) 323.427
Synonym N-[2-[(1R,2R)-3-Oxo-2-(2Z)-2-penten-1-ylcyclopentyl]acetyl]-L-isoleucine;Jasmonic Acid L-Isoleucine Conjugate;Jasmonic Acid Isoleucine Conjugate
IUPAC-namn (2S,3S)-3-methyl-2-[[2-[(1R,2R)-3-oxo-2-[(Z)-pent-2-enyl]cyclopentyl]acetyl]amino]pentanoic acid
LEDER CC\C=C/C[C@@H]1[C@@H](CC(=O)N[C@@H]([C@@H](C)CC)C(=O)O)CCC1=O
Specifikationer
Kemiskt namn eller material N-[(-)-Jasmonoyl]-(L)-isoleucine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C18H29NO4/c1-4-6-7-8-14-13(9-10-15(14)20)11-16(21)19-17(18(22)23)12(3)5-2/h6-7,12-14,17H,4-5,8-11H2,1-3H3,(H,19,21)(H,22,23)/b7-6-/t12-,13+,14+,17-/m0/s1
Kvantitet 50 mg
Formel vikt 323.21
Procent renhet >95%