Läs mer
N-Lauroyl-L-glutamic Acid (>80%), TRC
Produktkod. 30472123
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30472123 1 g
1 options
Produktkod. 30472123 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3001151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Dodecanoyl-L-glutamic Acid
CAS 3397-65-7
Molekylformel C17 H31 N O5
Kvantitet 1 g
Synonym N-(1-Oxododecyl)-L-glutamic Acid, (2S)-2-(Dodecanoylamino)pentanedioic Acid
InChI formel InChI=1S/C17H31NO5/c1-2-3-4-5-6-7-8-9-10-11-15(19)18-14(17(22)23)12-13-16(20)21/h14H,2-13H2,1H3,(H,18,19)(H,20,21)(H,22,23)/t14-/m0/s1
LEDER CCCCCCCCCCCC(=O)N[C@@H](CCC(=O)O)C(=O)O
IUPAC-namn (2S)-2-(dodecanoylamino)pentanedioic acid
Molekylvikt (g/mol) 329.432
Formel vikt 329.22