Läs mer
N-Lauroyl-N-methyltaurine Sodium Salt, TRC
Produktkod. 30180130
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180130 100 mg
30179911 1 g
30180406 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180130 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL185010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4337-75-1
Molekylformel C15H31NNaO4S
Molekylvikt (g/mol) 344.47
Synonym 2-[Methyl(1-oxododecyl)amino]-Ethanesulfonic Acid, Sodium Salt (9CI), Taurine, N-Lauroyl-N-methyl-, Sodium Salt (6CI,7CI,8CI), Diapon F, Diapon L, Hostapon LT Paste J, Hostapon LT Paste, Hostapon LT-SF, LMT, N-Dodecanoyl-N-methyltaurine Sodium Salt, N-Lauroyl-Nmethyltaurine Sodium Salt, N-Methyl-N-(2-sulfoethyl)lauramide Sodium Salt, Nikkol LMT, Nikkol LMT 30, Nikkol LMT-P, Nissan Diapon F, Nissan Diapon L, Sodium N-dodecanoyl-N-methyltaurinate, Sodium N-lauroyl-N-methylaminoethanesulfonate, Sodium N-lauroyl-N-methyltaurate, Sodium N-lauroyl-Nmethyltaurine, Sodium N-lauroylmethyltaurine, Sodium Methyltaurine Laurate
IUPAC-namn sodium;2-[dodecanoyl(methyl)amino]ethanesulfonate
LEDER FC(C=C1)=CC=C1C(CC[C@@H]2C(N(C3=CC=C(F)C=C3)[C@H]2C4=CC=C(O)C=C4)=O)=O
Specifikationer
Kemiskt namn eller material N-Lauroyl-N-methyltaurine Sodium Salt
Kvantitet 100 mg
InChI formel InChI=1S/C15H31NO4S.Na/c1-3-4-5-6-7-8-9-10-11-12-15(17)16(2)13-14-21(18,19)20;/h3-14H2,1-2H3,(H,18,19,20);/q;+1/p-1
Formel vikt 343.1793