Läs mer
N-Methoxysuccinyl-Ala-Ala-Pro-Val p-Nitroanilide, TRC
Produktkod. 30475263
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30475263 50 mg
1 options
Produktkod. 30475263 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26830050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methoxysuccinyl-Ala-Ala-Pro-Val p-Nitroanilide
CAS 70967-90-7
Molekylformel C27H38N6O9
Kvantitet 50 mg
Synonym MeOSuc-Ala-Ala-Pro-Val-pNA
InChI formel InChI=1S/C27H38N6O9/c1-15(2)23(26(38)30-18-8-10-19(11-9-18)33(40)41)31-25(37)20-7-6-14-32(20)27(39)17(4)29-24(36)16(3)28-21(34)12-13-22(35)42-5/h8-11,15-17,20,23H,6-7,12-14H2,1-5H3,(H,28,34)(H,29,36)(H,30,38)(H,31,37)/t16-,17-,20-,23-/m0/s1
LEDER COC(=O)CCC(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C(C)C)C(=O)Nc2ccc(cc2)[N+](=O)[O-]
IUPAC-namn methyl 4-[[(2S)-1-[[(2S)-1-[(2S)-2-[[(2S)-3-methyl-1-(4-nitroanilino)-1-oxobutan-2-yl]carbamoyl]pyrrolidin-1-yl]-1-oxopropan-2-yl]amino]-1-oxopropan-2-yl]amino]-4-oxobutanoate
Molekylvikt (g/mol) 590.63
Formel vikt 590.27