missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-[1-(thien-2-ylmethyl)piperid-4-yl]methylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | methyl({1-[(thiophen-2-yl)methyl]piperidin-4-yl}methyl)amine |
Formula Weight | 224.37 |
Percent Purity | 97% |
Chemical Name or Material | N-methyl-[1-(thien-2-ylmethyl)piperid-4-yl]methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'