missing translation for 'onlineSavingsMsg'
Läs mer

N-Methyl-2-phenylpropionamide, TRC

Produktkod. 30476129
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476129 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30476129 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30372010G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material N-Methyl-2-phenylpropionamide
CAS 58265-30-8
Molekylformel C10H13NO
Kvantitet 10 g
Synonym N,α-Dimethylbenzeneacetamide
InChI formel InChI=1S/C10H13NO/c1-8(10(12)11-2)9-6-4-3-5-7-9/h3-8H,1-2H3,(H,11,12)
LEDER CC(C1=CC=CC=C1)C(NC)=O
Molekylvikt (g/mol) 163.22
Formel vikt 163.1
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.