Läs mer
N-Methyl-3-pyrrolidinyl Cyclopentylmandelate (mixture of diastereomers), TRC
Produktkod. 30138246
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138246 100 mg
17983645 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138246 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM326040100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13118-11-1
Molekylformel C18 H25 N O3
Molekylvikt (g/mol) 303.4
Synonym Benzeneacetic acid, α-cyclopentyl-α-hydroxy-, 1-methyl-3-pyrrolidinyl ester, Mandelic acid, α-cyclopentyl-, 1-methyl-3-pyrrolidinyl ester (6CI,7CI), N-Methyl-3-pyrrolidinyl cyclopentylmandelate, 1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate, Glycopyrrolate Related Compound B as free base
IUPAC-namn (1-methylpyrrolidin-3-yl) 2-cyclopentyl-2-hydroxy-2-phenylacetate
LEDER CN1CCC(C1)OC(=O)C(O)(C2CCCC2)c3ccccc3
Specifikationer
Kemiskt namn eller material 1-Methylpyrrolidin-3-yl 2-Cyclopentyl-2-hydroxy-2-phenylacetate
Kvantitet 100 mg
InChI formel InChI=1S/C18H25NO3/c1-19-12-11-16(13-19)22-17(20)18(21,15-9-5-6-10-15)14-7-3-2-4-8-14/h2-4,7-8,15-16,21H,5-6,9-13H2,1H3
Formel vikt 303.1834