missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™

Catalog Number p-3486377
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
884507-32-8
C11H13N3O
203.25
MFCD09025868
MUCOLQGVWQXQMX-UHFFFAOYSA-N
n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, methyl 4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, benzenemethanamine,n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl, methyl 4-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
18525856
N-methyl-1-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
CNCC1=CC=C(C=C1)C1=NC(C)=NO1
This item is not returnable. View return policy
884507-32-8
C11H13N3O
203.25
MFCD09025868
MUCOLQGVWQXQMX-UHFFFAOYSA-N
n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, methyl 4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, benzenemethanamine,n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl, methyl 4-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
18525856
N-methyl-1-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
CNCC1=CC=C(C=C1)C1=NC(C)=NO1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl-[4-(3-methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'