Läs mer
N-Methyl-4-[4-[[[[3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-2-pyridinecarboxamide, TRC
Produktkod. 30473415
Change view
Kvantitet:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473415 1 g
30467139 100 mg
30478914 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30473415 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3313901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1285533-84-7
Molekylformel C21 H17 F3 N4 O3
Molekylvikt (g/mol) 430.38
Synonym 2-Pyridinecarboxamide, N-methyl-4-[4-[[[[3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-, N-Methyl-4-[4-[[3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]pyridine-2-carboxamide, Deschloro Sorafenib, N-Methyl-4-[4-[[[3-(trifluoromethyl)phenyl]carbamoyl]-amino]phenoxy]pyridine-2-carboxamide Imp. F (EP), 2-Pyridinecarboxamide, N-methyl-4-[4-[[[[3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-, N-Methyl-4-[4-[[3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]pyridine-2-carboxamide, Deschloro Sorafenib
IUPAC-namn N-methyl-4-[4-[[3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]pyridine-2-carboxamide
LEDER CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3cccc(c3)C(F)(F)F)cc2)ccn1
Specifikationer
Kemiskt namn eller material N-Methyl-4-[4-[([(3-(trifluoromethyl)phenyl)amino]carbonyl)amino]phenoxy]-2-pyridinecarboxamide
Kvantitet 1 g
InChI formel InChI=1S/C21H17F3N4O3/c1-25-19(29)18-12-17(9-10-26-18)31-16-7-5-14(6-8-16)27-20(30)28-15-4-2-3-13(11-15)21(22,23)24/h2-12H,1H3,(H,25,29)(H2,27,28,30)
Formel vikt 430.1253