Läs mer
N-Methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-3-carboxamide Hydrochloride, TRC
Produktkod. 30475495
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475495 100 mg
30474930 50 mg
30474518 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM334248100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1219957-27-3
Molekylformel C8H12N4O . HCl
Molekylvikt (g/mol) 216.67
Synonym 1H-Pyrazolo[4,3-c]pyridine-3-carboxamide, 4,5,6,7-tetrahydro-N-methyl-, hydrochloride (1:1) (ACI), N-Methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride, 4,5,6,7-Tetrahydro-N-methyl-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride, N-Methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
IUPAC-namn N-methyl-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide;hydrochloride
LEDER Cl.CNC(=O)c1n[nH]c2CCNCc12
Specifikationer
Kemiskt namn eller material N-Methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-3-carboxamide Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C8H12N4O.ClH/c1-9-8(13)7-5-4-10-3-2-6(5)11-12-7;/h10H,2-4H2,1H3,(H,9,13)(H,11,12);1H
Formel vikt 216.0778