Läs mer
N-Methyl-4-phenylpyridinium Iodide, TRC
Produktkod. 30005181
Change view
Kvantitet:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005181 1 g
30007127 25 mg
30006457 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005181 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3262851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36913-39-0
Molekylformel C12 H12 N . I
Molekylvikt (g/mol) 297.13
Synonym 1-Methyl-4-phenylpyridinium Iodide, MPP+ Iodide;
IUPAC-namn 1-methyl-4-phenylpyridin-1-ium;iodide
LEDER [I-].C[n+]1ccc(cc1)c2ccccc2
Specifikationer
Kemiskt namn eller material N-Methyl-4-phenylpyridinium Iodide
Kvantitet 1 g
InChI formel InChI=1S/C12H12N.HI/c1-13-9-7-12(8-10-13)11-5-3-2-4-6-11;/h2-10H,1H3;1H/q+1;/p-1
Formel vikt 297.0014