Läs mer
N-Methyl Amisulpride, TRC
Produktkod. 30007192
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007192 100 mg
30007165 50 mg
30006593 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007192 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM287320100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391054-22-0
Molekylformel C18 H29 N3 O4 S
Molekylvikt (g/mol) 383.51
Synonym N-Methylamisulpride, Benzamide, 4-amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxy-N-methyl-, 4-Amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxy-N-methyl-benzamide, 4-Amino-N-[[(2RS)-1-ethylpyrrolidin-2-yl]methyl]-5-(ethylsulfonyl)-2-methoxy-N-methylbenzamide
IUPAC-namn 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxy-N-methylbenzamide
LEDER CCN1CCCC1CN(C)C(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC
Specifikationer
Kemiskt namn eller material N-Methyl Amisulpride
Kvantitet 100 mg
InChI formel InChI=1S/C18H29N3O4S/c1-5-21-9-7-8-13(21)12-20(3)18(22)14-10-17(26(23,24)6-2)15(19)11-16(14)25-4/h10-11,13H,5-9,12,19H2,1-4H3
Formel vikt 383.1879