Läs mer
N-Methyl Anabasine, TRC
Produktkod. 30473838
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473838 25 mg
30469431 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30473838 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28750025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methyl Anabasine
CAS 2055-12-1
Anteckningar om renhetsgrad GC
Molekylformel C11 H16 N2
Kvantitet 25 mg
Synonym 1-Methyl-anabasine, 3-(1-Methyl-2-piperidinyl)-pyridine, 1-Methyl-2-(3'-pyridyl)piperidine, 1-Methylanabasine, N'-Methylanabasine, N'-Methylanabasine
InChI formel InChI=1S/C11H16N2/c1-13-8-3-2-6-11(13)10-5-4-7-12-9-10/h4-5,7,9,11H,2-3,6,8H2,1H3
LEDER CN1CCCCC1c2cccnc2
IUPAC-namn 3-(1-methylpiperidin-2-yl)pyridine
Molekylvikt (g/mol) 176.258
Formel vikt 176.131