Läs mer
N-(Methyl-d3) Trimetazidine Dihydrochloride, TRC
Produktkod. 30472321
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30472321 2.5 mg
1 options
Produktkod. 30472321 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3310432.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(Methyl-d3) Trimetazidine Dihydrochloride
CAS 1794760-10-3
Molekylformel C15 D3 H21 N2 O3
Kvantitet 2.5 mg
Synonym 1-(Methyl-d3)-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine Hydrochloride
InChI formel InChI=1S/C15H24N2O3/c1-16-7-9-17(10-8-16)11-12-5-6-13(18-2)15(20-4)14(12)19-3/h5-6H,7-11H2,1-4H3/i1D3
LEDER [2H]C([2H])([2H])N1CCN(Cc2ccc(OC)c(OC)c2OC)CC1
IUPAC-namn 1-(trideuteriomethyl)-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine
Molekylvikt (g/mol) 283.381
Formel vikt 283.198