Läs mer
N-Methyl DL-Pipecolic Acid, TRC
Produktkod. 30475648
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30475648 100 mg
1 options
Produktkod. 30475648 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM326345100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methyl DL-Pipecolic Acid
CAS 7730-87-2
Molekylformel C7 H13 N O2
Kvantitet 100 mg
Synonym 2-Piperidinecarboxylic acid, 1-methyl-, Pipecolic acid, 1-methyl- (6CI,7CI,8CI), 1-Methyl-2-piperidinecarboxylic Acid, 1-Methylpipecolinic acid, 2-Piperidinecarboxylic acid, 1-methyl-, (±)-, N-Methyl-2-piperidinecarboxylic acid
InChI formel InChI=1S/C7H13NO2/c1-8-5-3-2-4-6(8)7(9)10/h6H,2-5H2,1H3,(H,9,10)
LEDER CN1CCCCC1C(=O)O
IUPAC-namn 1-methylpiperidine-2-carboxylic acid
Molekylvikt (g/mol) 143.18
Formel vikt 143.0946