Läs mer
N-Methyl Formoterol Fumarate (2:1) (Mixture of Diastereomers), TRC
Produktkod. 30483225
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30483225 2.5 mg
1 options
Produktkod. 30483225 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2959502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methyl Formoterol Fumarate (2:1) (Mixture of Diastereomers)
CAS 1795133-97-9
Molekylformel 2 C20 H26 N2 O4 . C4 H4 O4
Kvantitet 2.5 mg
Synonym N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]methylamino]ethyl]phenyl]formamide 2-butenedioate (2:1), N-[2-Hydroxy-5-[1-hydroxy-2-[methyl-[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]formamide hemifumarate, N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]methylamino]ethyl]phenyl]formamide hemifumarate, N-[2-Hydroxy-5-[1-hydroxy-2-[[1-(4-methoxyphenyl)propan-2-yl](methyl)amino]ethyl]phenyl]formamide fumarate (2:1), N-Methyl Formoterol hemifumarate, N-Methylformoterol hemifumarate, N-[2-Hydroxy-5-[1-hydroxy-2-[methyl[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]formamide (2E)-2-Butenedioate Salt (2:1)
InChI formel InChI=1S/2C20H26N2O4.C4H4O4/c2*1-14(10-15-4-7-17(26-3)8-5-15)22(2)12-20(25)16-6-9-19(24)18(11-16)21-13-23;5-3(6)1-2-4(7)8/h2*4-9,11,13-14,20,24-25H,10,12H2,1-3H3,(H,21,23);1-2H,(H,5,6)(H,7,8)/b;;2-1+
LEDER COc1ccc(CC(C)N(C)CC(O)c2ccc(O)c(NC=O)c2)cc1.COc3ccc(CC(C)N(C)CC(O)c4ccc(O)c(NC=O)c4)cc3.OC(=O)\C=C\C(=O)O
IUPAC-namn (E)-but-2-enedioic acid;N-[2-hydroxy-5-[1-hydroxy-2-[1-(4-methoxyphenyl)propan-2-yl-methylamino]ethyl]phenyl]formamide
Molekylvikt (g/mol) 832.935
Formel vikt 832.3895