Läs mer
N-Methyl-L-alanine, TRC
Produktkod. 30483034
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30483034 10 g
1 options
Produktkod. 30483034 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM27656510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methyl-L-alanine
CAS 3913-67-5
Molekylformel C4H9NO2
Kvantitet 10 g
Synonym (S)-2-(Methylamino)propanoic Acid, (S)-N-Methylalanine, 2-(Methylamino)propanoic Acid, N-Methylalanine, alpha-(Methylamino)propionic Acid
InChI formel InChI=1S/C4H9NO2/c1-3(5-2)4(6)7/h3,5H,1-2H3,(H,6,7)/t3-/m0/s1
LEDER CN[C@@H](C)C(=O)O
IUPAC-namn (2S)-2-(methylamino)propanoic acid
Molekylvikt (g/mol) 103.12
Formel vikt 103.0633