Läs mer
N-Methyl Maprotiline Hydrochloride, TRC
Produktkod. 30472189
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30472189 50 mg
1 options
Produktkod. 30472189 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM18800550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine Hydrochloride
CAS 23941-45-9
Molekylformel C21 H25 N . Cl H
Kvantitet 50 mg
Synonym 9,10-Ethanoanthracene-9(10H)-propylamine, N,N-dimethyl-, hydrochloride (7CI,8CI), N-Methyl Maprotiline hydrochloride, N-Methylmaprotiline hydrochloride, 3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine hydrochloride
InChI formel InChI=1S/C21H25N.ClH/c1-22(2)15-7-13-21-14-12-16(17-8-3-5-10-19(17)21)18-9-4-6-11-20(18)21;/h3-6,8-11,16H,7,12-15H2,1-2H3;1H
LEDER Cl.CN(C)CCCC12CCC(c3ccccc13)c4ccccc24
IUPAC-namn 3-(9,10-Ethanoanthracen-9(10H)-yl)-N,N-dimethylpropan-1-amine hydrochloride
Molekylvikt (g/mol) 327.89
Formel vikt 327.1754