Läs mer
N-Methyl Mitomycin A, TRC
Produktkod. 30473159
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30473159 25 mg
1 options
Produktkod. 30473159 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32308025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methyl Mitomycin A
CAS 18209-14-8
Molekylformel C17H21N3O6
Kvantitet 25 mg
Synonym (1aS,8S,8aR,8bS)-8-[[(Aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-1,5-dimethylazirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, [1aS-(1aα,8β,8aα,8bα)]-8-[[(Aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-1,5-dimethylazirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, Mitomycin F, N-Methylmitomycin A, 1,1a,2,8,8a,8b-Hexahydro-8-(hydroxymethyl)-6,8aα-dimethoxy-1,5-dimethylazirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione Carbamate
InChI formel InChI=1S/C17H21N3O6/c1-7-12(21)11-10(13(22)14(7)24-3)8(6-26-16(18)23)17(25-4)15-9(19(15)2)5-20(11)17/h8-9,15H,5-6H2,1-4H3,(H2,18,23)/t8-,9+,15+,17-,19?/m1/s1
LEDER COC1=C(C)C(=O)C2=C([C@@H](COC(=O)N)[C@@]3(OC)[C@@H]4[C@H](CN23)N4C)C1=O
Molekylvikt (g/mol) 363.37