Läs mer
N-Methyl Moxifloxacin Hydrochloride, TRC
Produktkod. 30472779
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30472779 5 mg
1 options
Produktkod. 30472779 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3203355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methylmoxifloxacin Hydrochloride
CAS 1350716-67-4
Molekylformel C22 H26 F N3 O4 . Cl H
Kvantitet 5 mg
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-1-methyl-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Hydrochloride
InChI formel InChI=1S/C22H26FN3O4.ClH/c1-24-7-3-4-12-9-25(11-17(12)24)19-16(23)8-14-18(21(19)30-2)26(13-5-6-13)10-15(20(14)27)22(28)29;/h8,10,12-13,17H,3-7,9,11H2,1-2H3,(H,28,29);1H/t12-,17+;/m0./s1
LEDER Cl.COc1c(N2C[C@@H]3CCCN(C)[C@@H]3C2)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(=O)O
IUPAC-namn 7-[(4aS,7aS)-1-methyl-3,4,4a,5,7,7a-hexahydro-2H-pyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride
Molekylvikt (g/mol) 451.919
Formel vikt 451.167