missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(1-methyl-1H-indol-6-yl)methyl]amine, 97%, Thermo Scientific™

Catalog Number p-3486388
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
884507-20-4
C11H14N2
174.247
MFCD08690256
ODIDITAYVOGYIV-UHFFFAOYSA-N
n-methyl-n-1-methyl-1h-indol-6-yl methyl amine, methyl 1-methylindol-6-yl methyl amine, 1h-indole-6-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-6-yl methanamine, 1h-indole-6-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-6-ylmethyl-amine
18525770
N-methyl-1-(1-methylindol-6-yl)methanamine
CNCC1=CC2=C(C=C1)C=CN2C
This item is not returnable. View return policy
884507-20-4
C11H14N2
174.247
MFCD08690256
ODIDITAYVOGYIV-UHFFFAOYSA-N
n-methyl-n-1-methyl-1h-indol-6-yl methyl amine, methyl 1-methylindol-6-yl methyl amine, 1h-indole-6-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-6-yl methanamine, 1h-indole-6-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-6-ylmethyl-amine
18525770
N-methyl-1-(1-methylindol-6-yl)methanamine
CNCC1=CC2=C(C=C1)C=CN2C
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 174.25
Percent Purity 95%
Chemical Name or Material N-methyl-N-[(1-methyl-1H-indol-6-yl)methyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'