missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-n-(3-piperidin-1-ylbenzyl)amine, 90%, Thermo Scientific™

Catalog Number p-3175710
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
859850-65-0
C13H20N2
204.317
MFCD07772817
WHELHMLNRPDTJU-UHFFFAOYSA-N
n-methyl-n-3-piperidin-1-ylbenzyl amine, methyl 3-piperidin-1-yl phenyl methyl amine, methyl 3-piperidylphenyl methyl amine, benzenemethanamine,n-methyl-3-1-piperidinyl, n-methyl-1-3-piperidin-1-ylphenyl methanamine, n-methyl-1-3-piperidin-1-yl phenyl methanamine
7162073
N-methyl-1-(3-piperidin-1-ylphenyl)methanamine
CNCC1=CC(=CC=C1)N2CCCCC2
This item is not returnable. View return policy
859850-65-0
C13H20N2
204.317
MFCD07772817
WHELHMLNRPDTJU-UHFFFAOYSA-N
n-methyl-n-3-piperidin-1-ylbenzyl amine, methyl 3-piperidin-1-yl phenyl methyl amine, methyl 3-piperidylphenyl methyl amine, benzenemethanamine,n-methyl-3-1-piperidinyl, n-methyl-1-3-piperidin-1-ylphenyl methanamine, n-methyl-1-3-piperidin-1-yl phenyl methanamine
7162073
N-methyl-1-(3-piperidin-1-ylphenyl)methanamine
CNCC1=CC(=CC=C1)N2CCCCC2
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.32
Percent Purity 95%
Chemical Name or Material N-methyl-N-(3-piperidin-1-ylbenzyl)amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'