missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(6-pyrrolidin-1-ylpyridin-2-yl)methyl]amine, 97%, Thermo Scientific™

Catalog Number p-3522606
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
884507-14-6
C11H17N3
191.278
MFCD09025832
GGQYFTXRIMPZQO-UHFFFAOYSA-N
n-methyl-n-6-pyrrolidin-1-ylpyridin-2-yl methyl amine, 2-pyridinemethanamine,n-methyl-6-1-pyrrolidinyl, n-methyl 6-pyrrolidin-1-yl pyridin-2-yl methanamine, methyl 6-pyrrolidin-1-yl pyridin-2-yl methyl amine, methyl 6-pyrrolidinyl 2-pyridyl methyl amine, 2-pyridinemethanamine, n-methyl-6-1-pyrrolidinyl, n-methyl-1-6-1-pyrrolidinyl-2-pyridinyl methanamine, n-methyl-1-6-pyrrolidin-1-yl pyridin-2-yl methanamine
24229485
N-methyl-1-(6-pyrrolidin-1-ylpyridin-2-yl)methanamine
CNCC1=NC(=CC=C1)N2CCCC2
This item is not returnable. View return policy
884507-14-6
C11H17N3
191.278
MFCD09025832
GGQYFTXRIMPZQO-UHFFFAOYSA-N
n-methyl-n-6-pyrrolidin-1-ylpyridin-2-yl methyl amine, 2-pyridinemethanamine,n-methyl-6-1-pyrrolidinyl, n-methyl 6-pyrrolidin-1-yl pyridin-2-yl methanamine, methyl 6-pyrrolidin-1-yl pyridin-2-yl methyl amine, methyl 6-pyrrolidinyl 2-pyridyl methyl amine, 2-pyridinemethanamine, n-methyl-6-1-pyrrolidinyl, n-methyl-1-6-1-pyrrolidinyl-2-pyridinyl methanamine, n-methyl-1-6-pyrrolidin-1-yl pyridin-2-yl methanamine
24229485
N-methyl-1-(6-pyrrolidin-1-ylpyridin-2-yl)methanamine
CNCC1=NC(=CC=C1)N2CCCC2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material N-Methyl-N-[(6-pyrrolidin-1-ylpyridin-2-yl)methyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'