Läs mer
N'-Methyl-N,N-diphenylurea, TRC
Produktkod. 30479619
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30479619 5 g
1 options
Produktkod. 30479619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3037405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methyl-1,1-diphenylurea
CAS 13114-72-2
Anteckningar om renhetsgrad HPLC
Molekylformel C14 H14 N2 O
Kvantitet 5 g
Synonym Temozolomide Related Compound B, N'-Methyl-N,N-diphenylurea, Urea','3-methyl-1,1-diphenyl-, 3-Methyl-1,1-diphenyl-urea, Acardit II, Acardite II, Akardit II, Akardite II, N-Methyl-N',N'-diphenylurea
InChI formel InChI=1S/C14H14N2O/c1-15-14(17)16(12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11H,1H3,(H,15,17)
LEDER CNC(=O)N(c1ccccc1)c2ccccc2
IUPAC-namn 3-methyl-1,1-diphenylurea
Molekylvikt (g/mol) 226.27
Formel vikt 226.1106