Läs mer
N-Methyl-N-nitrosourethane, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Methyl-N-nitrosourethane |
| CAS | 615-53-2 |
| Molekylformel | C4 H8 N2 O3 |
| Kvantitet | 1 g |
| Synonym | Methylnitrosocarbamic Acid Ethyl Ester, Ethyl N-Methylnitrosocarbamate, Methylnitrosourethane, Nitrosomethylurethane, NMU, NSC 2860 |
| InChI formel | InChI=1S/C4H8N2O3/c1-3-9-4(7)6(2)5-8/h3H2,1-2H3 |
| LEDER | CCOC(=O)N(C)N=O |
| IUPAC-namn | ethyl N-methyl-N-nitrosocarbamate |
| Molekylvikt (g/mol) | 132.1179 |
| Formel vikt | 132.0535 |