Läs mer
N-Methyl-N-vinylacetamide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Methyl-N-vinylacetamide |
| CAS | 3195-78-6 |
| Molekylformel | C5 H9 N O |
| Kvantitet | 500 mg |
| Synonym | Acetamide, N-methyl-N-vinyl- (6CI,7CI,8CI), N-Ethenyl-N-methylacetamide, N-Methyl-N-vinylacetamide, N-Vinyl-N-methylacetamide |
| InChI formel | InChI=1S/C5H9NO/c1-4-6(3)5(2)7/h4H,1H2,2-3H3 |
| LEDER | CN(C=C)C(=O)C |
| IUPAC-namn | N-ethenyl-N-methylacetamide |
| Molekylvikt (g/mol) | 99.13 |
| Formel vikt | 99.0684 |