Läs mer
N-Methyl Paroxetine, TRC
Produktkod. 30005793
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005793 10 mg
30009866 50 mg
30009948 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005793 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32587010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 110429-36-2
Molekylformel C20 H22 F N O3
Molekylvikt (g/mol) 343.39
Synonym Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methyl-, (3S,4R)-, Piperidine, 3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methyl-, (3S-trans)-, (-)-trans-4-(4-Fluorophenyl)-3-(3,4-methylenedioxyphenoxymethyl)-1-methylpiperidine, N-Methylparoxetine
IUPAC-namn (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)-1-methylpiperidine
LEDER CN1CC[C@H]([C@H](COc2ccc3OCOc3c2)C1)c4ccc(F)cc4
Specifikationer
Kemiskt namn eller material trans(-)-1-Methyl-3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine
Kvantitet 10 mg
InChI formel InChI=1S/C20H22FNO3/c1-22-9-8-18(14-2-4-16(21)5-3-14)15(11-22)12-23-17-6-7-19-20(10-17)25-13-24-19/h2-7,10,15,18H,8-9,11-13H2,1H3/t15-,18-/m0/s1
Formel vikt 343.1584