Läs mer
N-Methyl-perfluorohexane-1-sulfonamide (>90%), TRC
Produktkod. 30467198
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30467198 250 mg
1 options
Produktkod. 30467198 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM327348250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-Methyl-perfluorohexane-1-sulfonamide
CAS 68259-15-4
Molekylformel C7 H4 F13 N O2 S
Kvantitet 250 mg
Synonym N-Methylperfluorohexane Sulfonamide, 1-Hexanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-N-methyl- (9CI, ACI), 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-N-methyl-1-hexanesulfonamide (ACI), N-Methyl-perfluorohexane-1-sulfonamide, N-Methylperfluorohexylsulfonamide
InChI formel InChI=1S/C7H4F13NO2S/c1-21-24(22,23)7(19,20)5(14,15)3(10,11)2(8,9)4(12,13)6(16,17)18/h21H,1H3
LEDER CNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluoro-N-methylhexane-1-sulfonamide
Molekylvikt (g/mol) 413.16
Formel vikt 412.9755