Läs mer
N-Methyl Topiramate, TRC
Produktkod. 30464973
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464973 100 mg
30479058 25 mg
30482967 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464973 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM330295100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 97240-80-7
Molekylformel C13H23NO8S
Molekylvikt (g/mol) 353.39
Synonym β-D-Fructopyranose, 2,3:4,5-bis-O-(1-methylethylidene)-, 1-(N-methylsulfamate), β-D-Fructopyranose, 2,3:4,5-bis-O-(1-methylethylidene)-, methylsulfamate (9CI), 5H-Bis[1,3]dioxolo[4,5-b:4',5'-d]pyran, β-D-fructopyranose deriv., N-Methyltopiramate, 2,3:4,5-bis-O-(1-Methylethylidene)-beta-D-Fructopyranose 1-(N-Methylsulfamate), 2,3:4,5-bis-O-(1-methylethylidene)-beta-D-fructopyranose Methylsulfamate, 5H-Bis[1,3]dioxolo[4,5-b:4',5'-d]pyran beta-D-Fructopyranose deriv.
IUPAC-namn [(3aS,5aR,8aR,8bS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[5,4-b:5',3'-d]pyran-3a-yl]methyl N-methylsulfamate
LEDER CNS(=O)(=O)OC[C@@]12OC[C@H]3OC(C)(C)O[C@H]3[C@@H]1OC(C)(C)O2
Specifikationer
Kemiskt namn eller material N-Methyl Topiramate
Kvantitet 100 mg
InChI formel InChI=1S/C13H23NO8S/c1-11(2)19-8-6-17-13(7-18-23(15,16)14-5)10(9(8)20-11)21-12(3,4)22-13/h8-10,14H,6-7H2,1-5H3/t8-,9-,10+,13+/m1/s1
Formel vikt 353.1144