missing translation for 'onlineSavingsMsg'
Läs mer

N-Methyl Trimetazidine-d8 Dihydrochloride, TRC

Product Code. 30473787
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30473787 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30473787 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM33104210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-Methyl Trimetazidine-d8 Dihydrochloride
CAS 1246819-10-2
Molekylformel C15 D8 H16 N2 O3
Kvantitet 10 mg
Synonym 1-Methyl-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine-d8 Hydrochloride
InChI formel InChI=1S/C15H24N2O3/c1-16-7-9-17(10-8-16)11-12-5-6-13(18-2)15(20-4)14(12)19-3/h5-6H,7-11H2,1-4H3/i7D2,8D2,9D2,10D2
LEDER [2H]C1([2H])N(C)C([2H])([2H])C([2H])([2H])N(Cc2ccc(OC)c(OC)c2OC)C1([2H])[2H]
IUPAC-namn 2,2,3,3,5,5,6,6-octadeuterio-1-methyl-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine
Molekylvikt (g/mol) 288.412
Formel vikt 288.229
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.