Läs mer
N-Methyl Trimetazidine Dihydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-Methyl-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride (N-Methyltrimetazidine Dihydrochloride) |
| CAS | 53960-20-6 |
| Molekylformel | C15 H24 N2 O3 . 2 Cl H |
| Kvantitet | 100 mg |
| Synonym | Piperazine, 1-methyl-4-[(2,3,4-trimethoxyphenyl)methyl]-, dihydrochloride (9CI), Piperazine, 1-methyl-4-(2,3,4-trimethoxybenzyl)-, dihydrochloride (7CI), 1-Methyl-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride, Trimetazidine Dihydrochloride Imp. I (EP) as Dihydrochloride, N-Methyltrimetazidine Dihydrochloride |
| InChI formel | InChI=1S/C15H24N2O3.2ClH/c1-16-7-9-17(10-8-16)11-12-5-6-13(18-2)15(20-4)14(12)19-3;;/h5-6H,7-11H2,1-4H3;2*1H |
| LEDER | Cl.Cl.COc1ccc(CN2CCN(C)CC2)c(OC)c1OC |
| IUPAC-namn | 1-methyl-4-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride |
| Molekylvikt (g/mol) | 353.28 |
| Formel vikt | 352.132 |